Ion 520™ & Ion 530™ Kit-OT2 - FAQs
Ion 520™ & Ion 530™ Kit-OT2 - FAQs
View additional product information for Ion 520™ & Ion 530™ Kit-OT2 - FAQs (A27751)
7 product FAQs found
What should I do if the centrifuge is still running even though the time remaining is 0:00 at the final spin of the Ion OneTouch 2?
The "time remaining" is not entirely accurate. The centrifuge may keep rotating for about 15 minutes after the time has become 0:00. If it rotates for more than 15 minutes, please contact Technical Support.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
What should I do if an "Over Pressure" error appears after I start an Ion OneTouch 2 run?
The pressure inside the instrument may be above normal. If the Ion OneTouch 2 firmware is older than v540, please update it to the latest version.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
What are these black precipitates in the recovery tube and waste of the Ion OneTouch System?
It may be rubber from the packing. The amount varies among instruments and it is not anything to be concerned about, as it will be removed by the Ion OneTouch ES.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
How do I update my Ion OneTouch 2 firmware?
Firmware updates to the software controlling the Ion OneTouch 2 Instrument are periodically released. To update the firmware to the appropriate version, use either a USB flash drive or an ethernet connection, as described on page 11 of the Ion OneTouch 2 System - Installation, Setup, and Maintenance User Guide (http://tools.thermofisher.com/content/sfs/manuals/MAN0014388_IonOneTouch2Sys_UG.pdf ).
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
What are the currently available Ion OneTouch 2 template kits?
For the Ion PGM System, we offer:
- Ion PGM Hi-Q View OT2 Kit (Cat. No. A29900) for template preparation of libraries up to 400 bp in length.
For the Ion Proton System, we offer:
- Ion PI Hi-Q OT2 200 Kit (Cat. No. A26434) for template preparation of libraries up to 200 bp in length.
For the Ion S5 System and Ion S5 XL System, we offer two kits:
- Ion 520/530 Kit-OT2 (Cat. No. A27751) for template preparation of libraries up to 400 bp in length.
- Ion 540 Kit- OT2 (Cat. No. A27753) for template preparation of libraries up to 200 bp in length.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
How much oil and recovery solution is typically consumed during a normal Ion Torrent Ion OneTouch 2 run and during a cleaning?
In a typical Ion OneTouch 2 run, you should expect ~18 mm drop in the level of oil in the tube (~22 mL consumed), and ~13 mm drop in the recovery solution (~15 mL consumed). During cleaning, you should expect to collect ~10 mL of oil in the 50 mL conical waste collection tube.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
Where can I find the protocols for my Ion Torrent Ion OneTouch 2 kit?
Ion Torrent OneTouch2 protocols can be found at our NGS Support Center (thermofisher.com/ngssupport), in the NGS Template Preparation Support Center section. For all other protocols, please go here (http://www.thermofisher.com/manuals-protocols-search.html).
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.