VetMAX™ African Swine Fever Virus Detection Kit
Applied Biosystems™
VetMAX™ African Swine Fever Virus Detection Kit
The VetMAX African Swine Fever Virus Detection Kit is a single-well duplex real-time PCR for the detection of African Swine Fever Virus (ASFV) DNA in serum, whole blood, and tissue samples of pigs using TaqMan primer and probes.
Have Questions?
Catalog NumberQuantity
A28809100 reactions
Catalog number A28809
Price (USD)
-
Quantity:
100 reactions
The VetMAX African Swine Fever Virus Detection Kit is a single-well duplex real-time PCR for the detection of African Swine Fever Virus (ASFV) DNA in serum, whole blood, and tissue samples of pigs using TaqMan primer and probes.
This kit enables detection in individual samples or pools of up to 10 samples.
The kit is sensitive and specific for the detection of ASFV DNA, allowing for reliable and rapid screening during outbreaks and monitoring situations. It has been validated by the European Union Reference Laboratory.The kit has been used since 2015 during outbreaks in Eastern Europe for clinical sign confirmation and detection of domestic and wild pigs affected by ASFV.
The kit includes:
  • A ready-to-use master mix for the detection of the ASFV target and the internal positive control
  • A positive control for ASFV
  • An internal positive control
For Veterinary Use Only. For In Vitro Use Only.
Specifications
Product LineVetMAX
Product TypeAfrican Swine Fever Virus Detection Kit
Quantity100 reactions
GC-Rich PCR PerformanceLow
PCR MethodqPCR
Unit SizeEach
Contents & Storage
Store at -10° to -30°C.