Oncomine™ Focus Assay, AmpliSeq™ Library, 48 Samples - FAQs
Oncomine™ Focus Assay, AmpliSeq™ Library, 48 Samples - FAQs
View additional product information for Oncomine™ Focus Assay, AmpliSeq™ Library - FAQs (A29230, A35957)
3 product FAQs found
How many samples can I process with the Oncomine Focus Assay, Chef-Ready Library?
The Oncomine Focus Assay, Chef-Ready Library contains the following components needed to prepare libraries for 32 samples:
- Oncomine Focus Assay, DNA Chef-Ready Panel (2X) - contains the DNA primer pool
- Oncomine Focus Assay, RNA Chef-Ready Panel (2X) - contains the RNA primer pool
- Ion AmpliSeq Kit for Chef Reagents DL8 cartridge - allows automation of the library prep on the Ion Chef Instrument
Note: The SuperScript transcriptase for cDNA synthesis is not included and must be purchased separately.
Does the Oncomine Focus Assay, AmpliSeq Library include reagents for template preparation and sequencing?
No, it only includes reagents for creating the library. For a complete kit, please see the Oncomine Focus Assay, 318 Solution, Cat. No. A28548. It includes all the necessary reagents and consumables for library construction, template preparation with the Ion OneTouch 2 System, and sequencing on the Ion PGM System.
What does the Oncomine Focus Assay, AmpliSeq Library, include?
Here are the components:
-Oncomine Focus Assay DNA Assay (48 reactions)
-Oncomine Focus Assay RNA Assay (48 reactions)
-Ion AmpliSeq Library Kit 2.0 (96 reactions)