Product Image
Ion Torrent™

Oncomine™ Focus Assay, AmpliSeq™ Library

The Oncomine Focus Assay is a targeted next-generation sequencing (NGS), multi-biomarker assay that enables the detection of variants across 52Read more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
A2923048 Samples
A3595716 Samples
Catalog number A29230
Price (USD)
-
Quantity:
48 Samples
The Oncomine Focus Assay is a targeted next-generation sequencing (NGS), multi-biomarker assay that enables the detection of variants across 52 cancer-relevant genes from DNA and RNA in a single workflow. The assay provides reagents for library construction and two pools—one DNA and one RNA—of multiplex PCR primers for preparation of amplicon libraries from formalin-fixed paraffin embedded (FFPE) tumor samples. Using as little as 10 ng of nucleic acid per primer pool, the assay enables analysis of hotpots, SNVs, indels, CNVs, and gene fusions in a single sequencing run.

The Oncomine Focus Assay, AmpliSeq Library, is for use as part of the Ion Chef and Ion GeneStudio S5 Systems workflow. Scalability and flexibility are achieved using Ion Xpress or IonCode barcode adapters for multiplexing barcoded samples on Ion S5 chips. For a kit that also includes reagents and consumables for automated library preparation on the Ion Chef System, please see the Oncomine Focus Assay, Chef-Ready Library.

From sample to answer
The Oncomine Focus Library Assay is part of an NGS workflow that helps researchers filter variant results from hundreds to just a few key cancer driver variants. Enabled by Oncomine informatics, this filtering, as well as the annotations from on-market labels, established guidelines (US-NCCN, US-FDA, EMSO, EMA), and global clinical trials, helps you contextualize and interpret findings to move toward the answers you need.

Learn more about Oncomine Focus Assay and Oncomine oncology.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Sequencing
For Use With (Equipment)Ion GeneStudio S5 System, Ion PGM™ System, Ion Chef™ System, Ion OneTouch™ 2 System
LibrariesTargeted Sequencing Library
No. of Samples48
Product LineAmpliSeq, Oncomine
Product TypeFocus Assay
Quantity48 Samples
Sample TypeDNA, RNA
Shipping ConditionDry Ice
Workflow StepLibrary Generation
Unit SizeEach
Package Components
A26120
Catalog Number
A26120
Unit Size
Qty
1
A28638
Catalog Number
A28638
Unit Size
Qty
1
Contents & Storage
• Oncomine Focus DNA Assay (48 reactions)
• Oncomine Focus RNA Assay (48 reactions)
• Ion AmpliSeq Library Kit 2.0 (96 reactions)
Store at -5 to -30°C.

• SuperScript for cDNA synthesis not included; must be purchased separately (Cat. No. A26123).
• Barcodes for multiplexing not included; must be purchased separately (Cat. Nos. A26119 or A29751).

Frequently asked questions (FAQs)

How many samples can I process with the Oncomine Focus Assay, Chef-Ready Library?

The Oncomine Focus Assay, Chef-Ready Library contains the following components needed to prepare libraries for 32 samples:

- Oncomine Focus Assay, DNA Chef-Ready Panel (2X) - contains the DNA primer pool
- Oncomine Focus Assay, RNA Chef-Ready Panel (2X) - contains the RNA primer pool
- Ion AmpliSeq Kit for Chef Reagents DL8 cartridge - allows automation of the library prep on the Ion Chef Instrument

Note: The SuperScript transcriptase for cDNA synthesis is not included and must be purchased separately.

Does the Oncomine Focus Assay, AmpliSeq Library include reagents for template preparation and sequencing?

No, it only includes reagents for creating the library. For a complete kit, please see the Oncomine Focus Assay, 318 Solution, Cat. No. A28548. It includes all the necessary reagents and consumables for library construction, template preparation with the Ion OneTouch 2 System, and sequencing on the Ion PGM System.

What does the Oncomine Focus Assay, AmpliSeq Library, include?

Here are the components:
-Oncomine Focus Assay DNA Assay (48 reactions)
-Oncomine Focus Assay RNA Assay (48 reactions)
-Ion AmpliSeq Library Kit 2.0 (96 reactions)