rhLaminin-521
Gibco™

rhLaminin-521

Recombinant human Laminin 521 (rhLaminin-521) is a recombinant human protein that provides a defined surface for feeder-free culture of humanRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
A29248100μg
A292491 mg
Catalog number A29248
Price (USD)
125.65
Online exclusive
131.00
Save 5.35
Each
Quantity:
100μg
Price (USD)
125.65
Online exclusive
131.00
Save 5.35
Each
Recombinant human Laminin 521 (rhLaminin-521) is a recombinant human protein that provides a defined surface for feeder-free culture of human pluripotent stem cells (PSCs). When used with Essential 8™ Medium, rhLaminin-521 has been proven to promote cellular survival under stressful conditions in the absence of small molecule inhibitors. rhLaminin-521 has been shown to support PSC growth for >20 passages without any signs of karyotypic abnormalities and to maintain the ability of PSCs to differentiate into all three germ line lineages. rhLaminin-521 also provides optimum reprogramming efficiency of human somatic cells as well as human pluripotent stem cell survival during feeder-dependent to feeder-free transitions when used in conjunction with Essential 8 Medium. For a kit that combines the medium and substrate, see the Essential 8™ Adaptation Kit.

rhLaminin-521 is:
• Robust—transition cells between feeder-dependent and feeder-free culture systems with minimal effort
• Efficient—easily reprogram human fibroblasts to pluripotent stem cells
• Physiologically relevant—expand cells with minimal differentiation
• Scalable—passage stem cell lines as single cells with no impact to genetic stability

Trouble-free transition
When used with Essential 8 Medium, rhLaminin-521 provides a robust culture matrix that supports pluripotent stem cell adherence and expansion during the transition from feeder-dependent culture systems.

Efficient reprogramming
rhLaminin-521 has been shown to increase the number of colonies obtained during reprogramming of human fibroblasts, which can be important for in vitro disease modeling.

Confident expansion
rhLaminin-521 is expressed in blastocysts and therefore provides an environment mimicking the human stem cell niche.

Easy scale-up
rhLaminin-521 promotes stable pluripotent stem cell maintenance as single cells in the absence of ROCK inhibitors.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell TypeStem Cells
Culture TypeStem Cell (Human, iPS - Induced Pluripotent Stem, Embryonic)
Purity or Quality GradehESC-Qualified
Quantity100μg
Shelf Life24 Months
Shipping ConditionDry Ice
SourceXeno-Free
ClassificationXeno-free
Product TypeExtracellular Matrix
pH6 to 8
Unit SizeEach
Contents & Storage
Store in freezer (-5 to -20°C) and protect from light.

Frequently asked questions (FAQs)

Can PSCs transitioned over into Essential 8 Medium on recombinant human laminin-521 (rhLaminin-521) from feeder-dependent cultures, then be transferred onto vitronectin (VTN-N)?

Yes. Following 2 passages on the rhLaminin-521 matrix, Versene or EDTA passaging should be used to subculture PSCs.

You recommend Geltrex matrix as the matrix for routine culture of StemFlex Medium. Can I also use Vitronectin or rhLaminin-521?

Yes. We recommend following the coating instructions for Vitronectin or rhLaminin-521 and using the culture and passaging recommendations in the StemFlex protocol.

I am interested in using StemFlex Medium in somatic cell reprogramming. What protocol should I follow for human fibroblasts?

We have evaluated the use of Cytotune-iPS 2.0 Sendai Reprogramming Kit (Cat. No. A16517) for somatic cell reprogramming of both neonatal and adult human dermal fibroblasts. For fibroblasts, follow the instructions provided in the Cytotune 2.0 reprogramming manual for feeder-free reprogramming (pgs. 16-20). On Day 7, you may use rhVTN-N (Cat. No. A400457 or A400458), Geltrex Flex matrix (Cat. No. A4000046801, A4000046802, or A4000046803), or rhLaminin-521 (Cat. No. A29248 or A29249). From Day 8 onward, rather than feeding daily with Essential 8 Medium, we recommend that you feed reprogrammed fibroblasts every-other-day with StemFlex Medium.

I am interested in using StemFlex Medium in somatic cell reprogramming. What protocol should I follow for blood cells?

We have evaluated the use of Cytotune-iPS 2.0 Sendai Reprogramming Kit (Cat. No. A16517) for somatic cell reprogramming of CD34+ blood cells. For CD34+ cells, follow the instructions provided in the Cytotune 2.0 reprogramming manual for feeder-free reprogramming (pgs. 39-44). On Day 3, you can utilize rhVTN-N (Cat. No. A400457 or A400458), Geltrex Flex matrix (Cat. No. A4000046801, A4000046802, or A4000046803), or rhLaminin-521 (Cat. No. A29248 or A29249). From Day 8 onward, rather than feeding daily with Essential 8 Medium, reprogrammed CD34+ cells should be fed every-other-day with StemFlex Medium.

What is the alternative for rhLaminin-521 (Cat. No. A29249)?

The appropriate alternative for rhLaminin-521 (Cat. No. A29249) would be either Laminin Mouse Protein, Natural (Cat. No. 23017015) or Vitronectin (VTN-N) Recombinant Human Protein, Truncated (Cat. No. A14700).

Find additional tips, troubleshooting help, and resources within our Stem Cell Culture Support Center.