The Applied Biosystems™ TaqMan™ Gene Expression Control Kit is designed to detect and quantify eukaryotic 18S ribosomal RNA (rRNA) andRead more
Have Questions?
Catalog Number
Quantity
A29349
250 reactions
Catalog number A29349
Price (USD)
399.65
Online exclusive
459.00
Save 59.35
Each
Quantity:
250 reactions
Price (USD)
399.65
Online exclusive
459.00
Save 59.35
Each
The Applied Biosystems™ TaqMan™ Gene Expression Control Kit is designed to detect and quantify eukaryotic 18S ribosomal RNA (rRNA) and human telomerase reverse transcriptase (hTERT) in human Raji cDNA on the QuantStudio™ 5 Real-Time PCR System. Gene-specific, ready-to-use TaqMan gene expression assays provide you with a fast, simple way to measure changes in gene expression in real-time PCR experiments.
Features of the TaqMan Gene Expression Control Kit include: • Simple—includes pre-formulated assays with our universal cycling conditions • Easy to use—just add to your real-time PCR experiment with no additional steps • Robust—generate reproducible quantitative gene expression data
Ready-to-use gene expression solution The TaqMan Gene Expression Control Kit contains TaqMan gene expression assays for eukaryotic 18S rRNA and hTERT and a sample of human Raji cDNA for use in TaqMan-based relative quantitation on the QuantStudio 5 Real-Time PCR System.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
FormatTube
Kit TypeGene Expression Control Kit
No. of Reactions250 Reactions
Product LineTaqMan
Quantity250 reactions
Shipping ConditionApproved for shipment on Wet or Dry Ice
SpeciesHuman
Target18 S rRNA & hTERT
For Use With (Application)Gene Expression
Label or DyeFAM
Unit SizeEach
Contents & Storage
Store in freezer (-5°C to -30°C).
Frequently asked questions (FAQs)
Do you offer a kit that contains TaqMan assays with positive control samples or templates?
Yes. The TaqMan Gene Expression Control Kit (Cat. No. A29349) contains TaqMan gene expression assays for eukaryotic 18S ribosomal RNA (rRNA) and human telomerase reverse transcriptase (hTERT) and a sample of human Raji cDNA for use in TaqMan assay-based relative quantitation.