TaqMan™ Gene Expression Control Kit - FAQs
TaqMan™ Gene Expression Control Kit - FAQs
View additional product information for TaqMan™ Gene Expression Control Kit - FAQs (A29349)
1 product FAQs found
Do you offer a kit that contains TaqMan assays with positive control samples or templates?
Yes. The TaqMan Gene Expression Control Kit (Cat. No. A29349) contains TaqMan gene expression assays for eukaryotic 18S ribosomal RNA (rRNA) and human telomerase reverse transcriptase (hTERT) and a sample of human Raji cDNA for use in TaqMan assay-based relative quantitation.