The Human Dopaminergic Neuron Immunocytochemistry Kit provides the NucBlue Fixed Cell Stain. If I don't have the correct filter set for this stain, can I use another nucleic acid stain?
If you use Alexa Fluor 488/Alexa Fluor 555 dye-conjugated secondary antibodies and you have a Cy5/Alexa Fluor 647 filter set, you may use a far-red nucleic acid stain, such as NucRed Dead 647 ReadyProbes Reagent (Cat. No. R37113) or SYTOX Deep Red Nucleic Acid Stain (Cat. No. S11380). Do not use these far-red nucleic acid stains if using an Alexa Fluor 594 dye-conjugated secondary antibody as there is too much spectral overlap of Alexa Fluor 594 into the Cy5/Alexa Fluor 647 range.