VetMAX™ Xeno™ Internal Positive Control - VIC™ Assay
The VetMAX™ Xeno™ Internal Positive Control (IPC) - VIC™ Assay is a primer-probe mix that detects the Xeno internal positiveRead more
Have Questions?
Change view
Catalog Number
No. of Reactions
A29765
100 Reactions
A29767
500 Reactions
2 Options
Catalog number A29765
Price (USD)
171.65
Online exclusive
191.00
Save 19.35
Each
No. of Reactions:
100 Reactions
Price (USD)
171.65
Online exclusive
191.00
Save 19.35
Each
The VetMAX™ Xeno™ Internal Positive Control (IPC) - VIC™ Assay is a primer-probe mix that detects the Xeno internal positive control. The resultant Xeno data is used to determine the validity of diagnostic test results. The Xeno IPC assay is introduced during the qPCR preparation step and carried through the animal health PCR workflow.
Features of the VetMAX Xeno IPC - VIC Assay include: • Provides confidence that qPCR test results are accurate and actionable • Easily integrates into any workflow • Greatly reduces the likelihood of false negatives
The VetMAX Xeno IPC - VIC Assay comes in a 25X concentration and easily integrates into animal health PCR workflows, regardless of the target assay, master mix, or sample preparation reagents already in place. Coupled with VetMAX™ Xeno Internal Positive Control RNA or VetMAX™ Xeno Internal Positive Control DNA, our proprietary design offers a verification layer to help ensure the qPCR test results are accurate and actionable by greatly reducing the likelihood of false negatives.
Xeno IPC assays are included in most VetMAX kits and have been successfully benchmarked against millions of genomes including those relevant to animal health. A VetMAX™ Xeno™ Internal Positive Control - LIZ™ Assay (Cat. No. A29766) is also available.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Fidelity (vs. Taq)25 X
FormatKit
No. of Reactions100 Reactions
Product LineVetMAX, Xeno
Product TypeInternal Positive Control Assay
Quantity100 reactions
Shipping ConditionDry Ice
SpeciesNot Species-Specific
Concentration25X
GC-Rich PCR PerformanceLow
Label or DyeVIC
PCR MethodqPCR
Unit SizeEach
Contents & Storage
1 x 110 μL Xeno VIC Primer Probe Mix. Store at -5°C to -30°C.