VetMAX™ Xeno™ Internal Positive Control - VIC™ Assay
The VetMAX™ Xeno™ Internal Positive Control (IPC) - VIC™ Assay is a primer-probe mix that detects the Xeno internal positiveRead more
Have Questions?
Change view
Catalog Number
No. of Reactions
A29767
500 Reactions
A29765
100 Reactions
2 Options
Catalog number A29767
Price (USD)
786.65
Online exclusive
874.00
Save 87.35
Each
No. of Reactions:
500 Reactions
Price (USD)
786.65
Online exclusive
874.00
Save 87.35
Each
The VetMAX™ Xeno™ Internal Positive Control (IPC) - VIC™ Assay is a primer-probe mix that detects the Xeno internal positive control. The resultant Xeno data is used to determine the validity of diagnostic test results. The Xeno IPC assay is introduced during the qPCR preparation step and carried through the animal health PCR workflow.
Features of the VetMAX Xeno IPC - VIC Assay include: • Provides confidence that qPCR test results are accurate and actionable • Easily integrates into any workflow • Greatly reduces the likelihood of false negatives
The VetMAX Xeno IPC - VIC Assay comes in a 25X concentration and easily integrates into animal health PCR workflows, regardless of the target assay, master mix, or sample preparation reagents already in place. Coupled with VetMAX™ Xeno Internal Positive Control RNA or VetMAX™ Xeno Internal Positive Control DNA, our proprietary design offers a verification layer to help ensure the qPCR test results are accurate and actionable by greatly reducing the likelihood of false negatives.
Xeno IPC assays are included in most VetMAX kits and have been successfully benchmarked against millions of genomes including those relevant to animal health. A VetMAX™ Xeno™ Internal Positive Control - LIZ™ Assay (Cat. No. A29766) is also available.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
FormatKit
No. of Reactions500 Reactions
Product LineVetMAX, Xeno
Product TypeInternal Positive Control
Quantity500 reactions
Shipping ConditionDry Ice
SpeciesNot Species-Specific
For Use With (Application)Animal Health
Label or DyeVIC
Unit SizeEach
Contents & Storage
1 x 550 μL Xeno VIC Primer Probe Mix. Store at -5°C to -30°C.