Alexa Fluor™ 790 NHS Ester (Succinimidyl Ester)
Invitrogen™

Alexa Fluor™ 790 NHS Ester (Succinimidyl Ester)

Alexa Fluor™ 790 is a bright and photostable near-IR dye that is spectrally similar to indocyanine green (ICG) and theRead more
Have Questions?
Catalog NumberQuantity
A30051100 μg
Catalog number A30051
Price (USD)
323.00
Each
Quantity:
100 μg
Request bulk or custom format
Price (USD)
323.00
Each
Alexa Fluor™ 790 is a bright and photostable near-IR dye that is spectrally similar to indocyanine green (ICG) and the IRDye™ 800 dye. Used for stable signal generation in imaging and flow cytometry, Alexa Fluor™ 790 dye is water soluble and pH-insensitive from pH 4 to pH 10. Fluorescence of this long-wavelength Alexa Fluor™ dye is not visible to the human eye but is readily detected by most imaging systems. As the longest-wavelength Alexa Fluor™ dye, the emission is well separated from commonly used far-red fluorophores such as Alexa Fluor™ 647 dye or allophycocyanin (APC), facilitating multicolor analysis. This fluorophore is also expected to be useful for small animal in-vivo imaging (SAIVI) applications or for two-color western applications with the LI-COR™ Odyssey™ infrared imaging system. In addition to reactive dye formulations, we offer Alexa Fluor™ 790 dye conjugated to a variety of antibodies, peptides, proteins, tracers, and amplification substrates optimized for cellular labeling and detection.The NHS ester (or succinimidyl ester) of Alexa Fluor™ 790 is the most popular tool for conjugating this dye to a protein or antibody. NHS esters can be used to label to the primary amines (R-NH2) of proteins, amine-modified oligonucleotides, and other amine-containing molecules. The resulting Alexa Fluor™ conjugate will exhibit brighter fluorescence and greater photostability than the conjugates of other spectrally similar fluorophores.

Detailed information about this AlexaFluor™ NHS ester:

Fluorophore label: Alexa Fluor™ 790 dye
Reactive group: NHS ester
Reactivity: Primary amines on proteins and ligands, amine-modified oligonucleotides
Ex/Em of the conjugate: 784/814 nm
Extinction coefficient: 260,000 cm-1M-1
Spectrally similar dyes: indocyanine green (ICG) and the IRDye™ 800 dye
Molecular weight: ∼1750

Typical Conjugation Reaction
You can conjugate amine-reactive reagents with virtually any protein or peptide (the provided protocol is optimized for IgG antibodies). You can scale the reaction for any amount of protein, but the concentration of the protein should be at least 2 mg/mL for optimal results. We recommend trying three different degrees of labeling, using three different molar ratios of the reactive reagent to protein.

The Alexa Fluor™ NHS ester is typically dissolved in high-quality anhydrous dimethylformamide (DMF) or dimethylsulfoxide (DMSO) (D12345), and the reaction is carried out in 0.1–0.2 M sodium bicarbonate buffer, pH 8.3, at room temperature for 1 hour. Because the pKa of the terminal amine is lower than that of the lysine epsilon-amino group, you may achieve more selective labeling of the amine terminus using a buffer closer to neutral pH.

Conjugate Purification
Labeled antibodies are typically separated from free Alexa Fluor™ dye using a gel filtration column, such as Sephadex™ G-25, BioGel™ P-30, or equivalent. For much larger or smaller proteins, select a gel filtration media with an appropriate molecular weight cut-off or purify by dialysis. We offer several purification kits optimized for different quantities of antibody conjugate:
Antibody Conjugate Purification Kit for 0.5-1 mg (A33086)
Antibody Conjugate Purification Kit for 20-50 μg (A33087)
Antibody Conjugate Purification kit for 50-100 μg (A33088)

Learn More About Protein and Antibody Labeling
We offer a wide selection of Molecular Probes™ antibody and protein labeling kits to fit your starting material and your experimental setup. See our Antibody Labeling kits or use our Labeling Chemistry Selection Tool for other choices. To learn more about our labeling kits, read Kits for Labeling Proteins and Nucleic Acids—Section 1.2 in The Molecular Probes™ Handbook.

We’ll Make a Custom Conjugate for You
If you can’t find what you’re looking for in our online catalog, we’ll prepare a custom antibody or protein conjugate for you. Our custom conjugation service is efficient and confidential, and we stand by the quality of our work. We are ISO 13485:2000 certified.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Chemical ReactivityAmine
Emission814 nm
Excitation784 nm
Label or DyeAlexa Fluor™ 790
Product TypeDye
Quantity100 μg
Reactive MoietyActive Ester, Succinimidyl Ester
Shipping ConditionRoom Temperature
Label TypeAlexa Fluor
Product LineAlexa Fluor
Unit SizeEach
Contents & Storage
Store in freezer (-5 to -30°C) and protect from light.

Frequently asked questions (FAQs)

I am labeling a protein with Alexa Fluor 488 SDP ester. The manual recommends using a sodium bicarbonate buffer at pH 8.3. Can I use a different buffer instead?

Yes. The important thing is to use a buffered solution with a pH between 8.0 and 8.5. Do not use Tris buffer, which has amine groups. Most other buffers will work fine in that pH range. This is also true for other amine-reactive dyes, such as succinimidyl (NHS) esters or TFP esters.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

I am not going to use all of my Alexa Fluor succinimidyl ester reactive dye. Can I just make it up in DMSO and store aliquots at -20 degrees C?

This is not recommended. Any trace amounts of water in the DMSO can promote spontaneous hydrolysis over time. Even if using anhydrous DMSO, DMSO is hygroscopic; it readily absorbs moisture from the atmosphere over time. A better alternative is to dissolve the reactive dye in a volatile solvent, make smaller aliquots and then evaporate off the solvent using a vacuum pump. The smaller aliquots of solid reactive dye should then be stored frozen, desiccated and protected from light. Contact Technical Support by sending an email to techsupport@thermofisher.com for the recommended volatile solvent.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

I am interested in purchasing Alexa Fluor 790 carboxylic acid derivative to be used as a free dye for a microscope standard. I see that you only offer Alexa Fluor 790 NHS Ester (Succinimidyl Ester) (Cat. No. A30051). Do you have any other recommendations?

We do not offer Alexa Fluor 790 carboxylic acid derivative. We only offer Alexa Fluor 790 NHS Ester (Cat. No. A30051). It is a succinimidyl ester (SE) derivative where the SE functional group (which makes the dye amine-reactive) is attached via a carboxylic acid functional group (thus the nomenclature).
Note: If the reactive moiety is an issue for your assay, then you can bring up the dye in an aqueous buffer, and leave it for about an hour, the SE will hydrolyze and become non-reactive.

Citations & References (2)

Citations & References
Abstract
Evaluation of fluorophores for optimal performance in localization-based super-resolution imaging.
Authors:Dempsey GT, Vaughan JC, Chen KH, Bates M, Zhuang X,
Journal:Nat Methods
PubMed ID:22056676
'One approach to super-resolution fluorescence imaging uses sequential activation and localization of individual fluorophores to achieve high spatial resolution. Essential to this technique is the choice of fluorescent probes; the properties of the probes, including photons per switching event, on-off duty cycle, photostability and number of switching cycles, largely dictate ... More
NHERF2 protein mobility rate is determined by a unique C-terminal domain that is also necessary for its regulation of NHE3 protein in OK cells.
Authors:Yang J, Singh V, Cha B, Chen TE, Sarker R, Murtazina R, Jin S, Zachos NC, Patterson GH, Tse CM, Kovbasnjuk O, Li X, Donowitz M,
Journal:J Biol Chem
PubMed ID:23612977
Na(+)/H(+) exchanger regulatory factor (NHERF) proteins are a family of PSD-95/Discs-large/ZO-1 (PDZ)-scaffolding proteins, three of which (NHERFs 1-3) are localized to the brush border in kidney and intestinal epithelial cells. All NHERF proteins are involved in anchoring membrane proteins that contain PDZ recognition motifs to form multiprotein signaling complexes. In ... More