qualyfast™ Legionella Plus
qualyfast™ Legionella Plus
qualyfast™ Legionella Plus
qualyfast™ Legionella Plus
Thermo Scientific™

qualyfast™ Legionella Plus

A NEW ERA IN qPCR ASSAYS

Pre-dosed and ready-to-use kit to measure Legionella spp., L. pneumophila, and L. pneumophila Serogroup 1 from the same tube.

Have Questions?
Catalog NumberQuantity
A30106696 tests
Catalog number A301066
Price (USD)
-
Quantity:
96 tests

qPCR detection kit for Legionella spp., L. pneumophila, and L. pneumophila SG1 with Inhibition Amplification Control.

The qualyfast™-plus Legionella test solution offers rapid, sensitive, specific, and reliable detection of Legionella species, L. pneumophila, and L. pneumophila Serogroup 1 from clean and dirty water samples.

  • Quick - Results in under 3 hours for rapid reporting
  • User-friendly - Room temperature stable to free-up cold storage space
  • Available in kits of 96 reactions
  • Multiplexed - Identification of Legionella species L. pneumophila, and L. pneumophila Serogroup 1 in a single PCR test
For Laboratory Use
Specifications
For Use With (Equipment)Applied Biosystems 7500 Fast Food Safety Real-Time PCR instrument and Applied Biosystems QuantStudio 5 Food Safety Real Time PCR System
No. of Reactions96
Product LineLegionella PCR and DNA Extraction Kit
Quantity96 tests
Shelf Life365 days
Sufficient For96 reactions
Unit SizeEach
Contents & Storage
Ambient