MicroAmp™ 8-Tube Strip with Attached Domed Caps, 0.2 mL - FAQs
MicroAmp™ 8-Tube Strip with Attached Domed Caps, 0.2 mL - FAQs
View additional product information for MicroAmp™ 8-Tube Strip with Attached Domed Caps, 0.2 mL - FAQs (A30589)
4 product FAQs found
What are the benefits of using MicroAmp 8-Tube Strip with Attached Domed Caps?
These new strips have new features including a graduated 20 µL measuring mark for visual verification and dual end tabs for better labeling and handling. In addition, the individual caps help prevent cross contamination and reduce sample evaporation.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.
Is the MicroAmp 8-Tube Strip with Attached Domed Caps DNase free?
Yes. The tubes are DNase/RNase/Human DNA free, as they are manufactured in a Class 100K ISO certified clean room UK production facility.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.
Do I need to use a tray with the MicroAmp 8-Tube Strip with Attached Domed Caps?
MicroAmp 96-Well Tray for VeriFlex Blocks (Cat. No. 4379983) is needed to load 1 or 2 MicroAmp Optical 8-Tube Strips with Attached Domed Caps on the ProFlex, SimpliAmp, MiniAmp, MiniAmp Plus, and Veriti Thermal Cycler 96-well blocks.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.
Which instrument(s) is the MicroAmp 8-Tube Strip with Attached Domed Caps compatible with?
The MicroAmp 8-Tube Strip with Attached Domed Caps is compatible with the following instruments:
- ProFlex 3 x 32-well PCR System
- ProFlex 96-well
- Veriti 96-well Thermal Cycler
- SimpliAmp Thermal Cycler
- MiniAmp Thermal Cycler
- MiniAmp Plus Thermal Cycler
- 2720 Thermal Cycler
- GeneAmp PCR System 9700, 96-well
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.