JetFlex™ Genomic DNA Purification Kit, 500 Preps - FAQs
JetFlex™ Genomic DNA Purification Kit, 500 Preps - FAQs
View additional product information for JetFlex™ Genomic DNA Purification Kit - FAQs (A30701)
1 product FAQs found
Does the JetFlex Genomic DNA Purification Kit work with frozen blood?
Yes. However, the yield from frozen blood is significantly lower than the yield from fresh blood.