PureLink™ Expi Endotoxin-Free Maxi Plasmid Purification Kit, 10 Preps - FAQs
PureLink™ Expi Endotoxin-Free Maxi Plasmid Purification Kit, 10 Preps - FAQs
View additional product information for PureLink™ Expi Endotoxin-Free Maxi Plasmid Purification Kit - FAQs (A31231, A31217, A33073)
1 product FAQs found
Do you offer a kit for plasmid extraction with endotoxin reduction?
We offer the following silica column-based plasmid purification kits for isolation of endotoxin-free or low-endotoxin plasmid DNA:
- PureLink Expi Endotoxin-Free Maxi Plasmid Purification Kit (Cat. No. A31217, A31231, A33073) (<0.1 EU/µg endotoxins):
https://www.thermofisher.com/order/catalog/product/A31217
- PureLink Fast Low-Endotoxin Midi (Cat. No. A35892, A36227) or Maxi (Cat. No. A36228, A35895) Plasmid Purification Kits (<1 EU/µg endotoxins):
https://www.thermofisher.com/order/catalog/product/A35892
https://www.thermofisher.com/order/catalog/product/A36228