PureLink™ Expi Endotoxin-Free Giga Plasmid Purification Kit - FAQs
PureLink™ Expi Endotoxin-Free Giga Plasmid Purification Kit - FAQs
View additional product information for PureLink™ Expi Endotoxin-Free Plasmid Purification Kits - FAQs (A31233, A31233-1, A31233-2, A31233-3, A31233-4, A31233-13, A31233-5, A31233-14, A31233-6, A31233-11, A31233-7, A31233-12, A31233-8, A31233-9, A31233-10)
3 product FAQs found
What makes the PureLink Expi Endotoxin-Free Mega/Giga plasmid purification kits yield endotoxin-free plasmid DNA?
The anion exchange resin used in PureLink HiPure plasmid purification kits produce plasmid DNA with inherently low endotoxin levels of 0.1-1.0 EU/µg. To add to this, PureLink Expi Endotoxin-Free Mega/Giga plasmid purification kits contain endotoxin-free components, and a proprietary Endotoxin Removal Buffer that further enhances endotoxin depletion, resulting in plasmid DNA that is consistently endotoxin-free (< 0.1 EU/µg) .
Is the Endotoxin Removal Buffer (ER) in PureLink Expi Endotoxin-Free Mega/Giga plasmid purfiication kits sold separately?
No. Endotoxin Removal Buffer (ER) in PureLink Expi Endotoxin-Free Mega/Giga plasmid purification kits is not sold separately.
What is the endotoxin level in the plasmid DNA prepared using PureLink Expi Endotoxin-Free Mega/Giga plasmid purfiication kits?
The PureLink Expi Endotoxin-Free Mega/Giga plasmid purification kits can produce endotoxin-free (< 0.1 EU/µg) advanced transfection-grade plasmid DNA.