Pentosidine (CAY-10010254-10mg) - FAQs
Pentosidine (CAY-10010254-10mg) - FAQs
View additional product information for Pentosidine (CAY-10010254-10mg) - FAQs (A32246)
1 product FAQs found
Can I store the MicroSEQ ID Ultra Sequencing Clean-up 8-Strips Kit (Cat. No. A33246) at room temperature (20-24 degrees C)?
Yes. The kit contents are stable when stored in their original packaging at room temperature for up to 4 weeks. However, the recommended storage condition is at 4 degrees C.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Applications Support Center.