Precision ID DL8 Kit - FAQs
Precision ID DL8 Kit - FAQs
View additional product information for Precision ID DL8 Kit - FAQs (A33212)
4 product FAQs found
How many Ion Chef runs can be performed with the Precision ID DL8 Kit?
The Precision ID DL8 Kit contains materials sufficient for performing 4 Ion Chef runs.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
What is the recommended amount of input DNA for the Precision ID DL8 Kit?
The recommended input DNA amount for the Precision ID DL8 Kit is 125 pg.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
How many reactions can be performed with the Precision ID DL8 Kit?
The Precision ID DL8 Kit provides reagents for 32 reactions.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
What is the Precision ID DL8 Kit (Cat. No. A33212)?
The Precision ID DL8 Kit is designed for automated library construction using the Precision ID GlobalFiler NGS STR Panel v2 on the Ion Chef System.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.