Pierce™ Intact Protein Standard Mix, 5 x 76 μg - FAQs
Pierce™ Intact Protein Standard Mix, 5 x 76 μg - FAQs
View additional product information for Pierce™ Intact Protein Standard Mix - FAQs (A33527, A33526)
1 product FAQs found
Which standards and calibrants do you recommend using for assessing LC-MS system suitability?
Here are our recommendations:
- Pierce Peptide Retention Time Calibration Mixture (Cat. No. 88321) for LC system and gradient optimization
- Pierce BSA Protein Digest, MS grade (Cat. No. 88341), Pierce 6 Protein Digest, equimolar, LC-MS grade (Cat. No. 88342), and Pierce HeLa Protein Digest Standard (Cat. Nos. 88328, 88329) for "bottom up" LC-MS method optimization and LC-MS suitability assessment
- Pierce LC-MS/MS System Suitability Standard (7 x 5 Mix) (Cat. No. A40010) for assessing the gradient, sensitivity, and dynamic range of the LC-MS system for discovery and targeted peptide quantitative workflows
- Pierce TMT11plex Yeast Digest Standard (Cat. Nos. A40938, A40939) for assessing system performance for TMT-based quantitative workflows
- Pierce Intact Protein Standard Mix (Cat. Nos. A33526, A33527) for "top down" LC-MS method optimization and LC-MS suitability assessment
Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.