B-27 Supplement, XenoFree, minus vitamin A, is a Research Use Only supplement based on the classic B-27 Supplement formulation and자세히 알아보기
Have Questions?
카탈로그 번호
수량
A3353501
10 mL
카탈로그 번호 A3353501
제품 가격(KRW)
409,000
Each
수량:
10 mL
제품 가격(KRW)
409,000
Each
B-27 Supplement, XenoFree, minus vitamin A, is a Research Use Only supplement based on the classic B-27 Supplement formulation and contains human-derived components and recombinant proteins.
Can I thaw B-27 Supplements in a 37 degrees C water bath or incubator?
We do not recommend thawing at 37 degrees C. Best results are observed upon thawing at room temperature for approximately 2 hrs or overnight at 2-8 degrees C.