B-27™ Supplement, XenoFree, minus vitamin A - FAQs
B-27™ Supplement, XenoFree, minus vitamin A - FAQs
View additional product information for B-27™ Supplement, XenoFree, minus vitamin A - FAQs (A3353501)
4 product FAQs found
Do you offer B-27 supplements with and without insulin?
Yes, we do offer B-27 Supplement, minus insulin (Cat. Nos. A1895601 and A1895602). All our other B-27 supplements do contain insulin.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Can I thaw B-27 Supplements in a 37 degrees C water bath or incubator?
We do not recommend thawing at 37 degrees C. Best results are observed upon thawing at room temperature for approximately 2 hrs or overnight at 2-8 degrees C.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
How do I thaw B-27 Supplements?
We recommend thawing it at 4 degrees C overnight or at room temperature for 2 hours. We don't recommend thawing in a water bath or incubator.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Can I re-freeze B-27 Supplements into smaller aliquots for future use?
Yes, we recommend that you aliquot when you first thaw it and thereafter subject the aliquot to no more than 2 additional freeze/thaw cycles.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.