Amplex™ Red/UltraRed Stop Reagent - FAQs
Amplex™ Red/UltraRed Stop Reagent - FAQs
View additional product information for Amplex™ Red/UltraRed Stop Reagent - FAQs (A33855)
1 product FAQs found
Can Amplex Red Assays be performed using cell lysates?
This is not recommended. The presence of endogenous proteases can complicate the assay by degrading the horseradish peroxidase (HRP). Endogenous peroxidases and antioxidants can modify the H2O2 required for the reaction, competing with HRP (and catalase) for the substrate.
The Amplex Red Assays are best performed with either purified enzymes or extracted H2O2 in a defined buffer system, extracellular solutions or body fluids (media, serum, etc.) that do not exhibit high levels of endogenous protease or oxidase activity and do not contain antioxidants.