SILAC Protein Quantitation Kit (Trypsin), DMEM - FAQs
SILAC Protein Quantitation Kit (Trypsin), DMEM - FAQs
View additional product information for SILAC Protein Quantitation Kit (Trypsin), DMEM - FAQs (A33972)
1 product FAQs found
Do you offer an alternative for SILAC Protein Quantitation Kit (Trypsin), DMEM (Cat. No. A33972)?
We do not offer an alternative for SILAC Protein Quantitation Kit (Trypsin), DMEM (Cat. No. A33972). We only have two kits specifically formulated for the SILAC Protein Quantitation Kit (Trypsin), which include the DMEM (Cat. No. A33972) and RPMI 1640 (Cat. No. A33973). We do offer the DMEM for SILAC (Cat. No. 88364), RPMI-1640 Medium for SILAC (Cat. No. 88370), DMEM:F12 for SILAC (Cat. No. 88370), and Dialyzed Fetal Bovine Serum (Cat. No. 26400036), as stand-alone products. However, we do not recommend replacing DMEM for SILAC with any other media, as each cell line has specific requirements for optimal growth.
A description of our SILAC Protein Quantitation Kits and kit components can be found in the User Guide.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.