Product Image
Applied Biosystems™

Automated Thermal Cycler (ATC) Block Module, 384 well

This 384-well Automated Thermal Cycler Block Module is a component of the 384-well Automated Thermal Cycler (ATC). An ATC ControlRead more
Have Questions?
Catalog NumberQuantity
A339761 module
Catalog number A33976
Price (USD)
12,630.00
Each
Quantity:
1 module
Price (USD)
12,630.00
Each
This 384-well Automated Thermal Cycler Block Module is a component of the 384-well Automated Thermal Cycler (ATC). An ATC Control Module is required for operation.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Depth (English)12.5 in.
Depth (Metric)31.7 cm
For Use With (Application)PCR
Height (English)5.3 in.
Height (Metric)13.3 cm
High-throughput CompatibilityDesigned for High-throughput Robotic Workflows
Max. Ramp Rate2.8°C/s (Block), 1.6°C/s (Sample)
Quantity1 module
Reaction SpeedStandard
Shipping ConditionApproved for Shipment at Room Temperature or on Wet Ice
Thermal Accuracy±0.25°C (35°C to 99.9°C)
Thermal Range4°C to 105°C
Thermal Uniformity<0.5°C (20 s after reaching 95°C)
Voltage100 to 240 V
WarrantyLimited Warranty
Wattage600 W
Weight6 kg (13.2 lb.)
Width (English)7.1 in.
Width (Metric)17.9 cm
Block Configurations384-well, 0.02 mL Block
Capacity1 x 384-well Full Skirted Plate
Product TypeThermal Cycler
Unit SizeEach

Frequently asked questions (FAQs)

What is the optimal ramp rate for my thermal cycler?

There is no universal ramp rate for a thermal cycler because both the block and sample ramp rates are different for each thermal cycler. The sample ramp rate changes with the volume of the sample since more time and energy are required to heat higher sample volumes. The ramp rate may also need to be changed for a particular PCR assay or primer pair if you are trying to use the same PCR assay on two different thermal cyclers, for example.

As a reference, the block ramp rate commonly used with DreamTaq Polymerase on the MiniAmp thermal cycler is 3.0 C/sec. In a 50 µL PCR reaction, this translates to a sample ramp rate of ~1 C/sec.

Please refer to the following application note for further information about thermocycler ramp rates: https://assets.thermofisher.com/TFS-Assets/LSG/brochures/Thermal_Cycler_Ramp_RatesAppNote.pdf

Find additional tips, troubleshooting help, and resources within ourPCR Thermal Cyclers Support Center.