Ion S5™ 2X Cleaning Solution
Thermo Scientific™

Ion S5™ 2X Cleaning Solution

Ion S5 2X Cleaning Solution is used to perform a pulse clean of your Ion S5 Sequencer to either recover the instrument performance from a diagnosed solid-state buffer leak or perform preventative maintenance if the instrument has not been used for several months.
Have Questions?
Catalog NumberQuantity
A341722 Bottles
Catalog number A34172
Price (USD)
396.00
Each
Quantity:
2 Bottles
Price (USD)
396.00
Each

Ion S5 2X Cleaning Solution is used to perform a pulse clean of your Ion S5 Sequencer to either recover the instrument performance from a diagnosed solid-state buffer leak or perform preventative maintenance if the instrument has not been used for several months.

Features include:

  • The ability to run a pulse clean in the event of a solid-state buffer leak
  • The option to deep clean the fluidics of the Ion S5 Sequencer
WARNING: Cancer – www.P65Warnings.ca.gov
Specifications
For Use With (Equipment)Ion GeneStudio S5 and S5
Height (Metric)131.1 mm
Length (Metric)61 mm
Product Type2X Cleaning Solution
Quantity2 Bottles
Width (Metric)61 mm
Unit SizeEach

Frequently asked questions (FAQs)

With Ion S5 2X Cleaning Solution, how long does a "pulse clean" take to complete?

It takes approximately 47 mins to do a "pulse clean",

When using Ion S5 2X Cleaning Solution, where in the software should I select "pulse clean"?

Go to "System tools" and select "pulse clean". The instrument must already have been cleaned after a run.

What are the dimensions of the bottles in Ion S5 2X Cleaning Solution?

Here are the bottle dimensions:
- Length (Metric) 61 mm
- Height (Metric) 131.1 mm
- Width (Metric) 61 mm

How many bottles of Ion S5 2X Cleaning Solution do you provide in Cat. No. A34172?

The kit contains 2 bottles of Ion S5 2X Cleaning Solution.

Which instruments is Ion S5 2X Cleaning Solution compatible with?

Ion S5 2X Cleaning Solution is compatible with GeneStudio S5/GeneStudio S5 Plus/GeneStudio S5 Prime systems and Ion S5/Ion S5 XL systems.