With Ion S5 2X Cleaning Solution, how long does a "pulse clean" take to complete?
It takes approximately 47 mins to do a "pulse clean",
When using Ion S5 2X Cleaning Solution, where in the software should I select "pulse clean"?
Go to "System tools" and select "pulse clean". The instrument must already have been cleaned after a run.
What are the dimensions of the bottles in Ion S5 2X Cleaning Solution?
Here are the bottle dimensions:
- Length (Metric) 61 mm
- Height (Metric) 131.1 mm
- Width (Metric) 61 mm
How many bottles of Ion S5 2X Cleaning Solution do you provide in Cat. No. A34172?
The kit contains 2 bottles of Ion S5 2X Cleaning Solution.
Which instruments is Ion S5 2X Cleaning Solution compatible with?
Ion S5 2X Cleaning Solution is compatible with GeneStudio S5/GeneStudio S5 Plus/GeneStudio S5 Prime systems and Ion S5/Ion S5 XL systems.
When should I use Ion S5 2X Cleaning Solution?
Ion S5 2X Cleaning Solution is used to perform a pulse clean of your Ion S5 Sequencer (GeneStudio S5/GeneStudio S5 Plus/GeneStudio S5 Prime systems and Ion S5/Ion S5 XL systems) to either recover the instrument performance from a diagnosed solid-state buffer leak or perform preventative maintenance if the instrument has not been used for several months.