본 제품은 액체 질소로 배송 및 보관되는 세포 관련 제품으로, 해당 제품 구매 시 세포 사용 가이드라인을 반드시 확인해주십시오. 세포사용가이드라인 다운로드
CF1 Mouse Embryonic Fibroblasts, irradiated
Gibco™
CF1 Mouse Embryonic Fibroblasts, irradiated
Irradiated Gibco CF1 Mouse Embryonic Fibroblasts (MEFs) are feeder cells that are ideal for supporting healthy undifferentiated human and mouse자세히 알아보기
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호수량
A341802 x 106 cells
A341814 x 106 cells
카탈로그 번호 A34180
제품 가격(KRW)
75,000
Each
수량:
2 x 106 cells
제품 가격(KRW)
75,000
Each
Irradiated Gibco CF1 Mouse Embryonic Fibroblasts (MEFs) are feeder cells that are ideal for supporting healthy undifferentiated human and mouse embryonic stem cells (ESCs) and induced pluripotent stem cells (iPSCs) in culture. Manufactured by MTI-GlobalStem and equivalent to MTI-GlobalStem Cat. Nos.GSC-6001G and GSC-6201G, these widely-used MEF cells are isolated from outbred CF1 mice, mitotically arrested by irradiation, and tested to ensure safety and performance. CF1 MEFs are traditional feeder cells for culturing pluripotent stem cells; for MEFs derived from ICR mice, see Mouse (ICR) Inactivated Embryonic Fibroblasts.
• Help save precious time with pre-inactivated, ready-to-use MEFs
• Culture ESCs and iPSCs with confidence using meticulously tested feeder cells
• Rely on a widely-used product
Help save time with ready-to-use MEFs
Irradiated CF1 MEFs help save time and trouble dealing with an animal facility, performing tedious, time-consuming dissections, and expanding and testing cells day after day, week after week. Focus on your research with these pre-inactivated, ready-to-use feeders.
Culture with meticulously tested feeder cells
Each lot of CF1 MEFs is tested for bacteria, fungi, mycoplasma, and mouse pathogens to minimize the risk of contamination. Each lot is also tested for post-thaw viability (typically greater than 95%) and ability to support mouse and human ES cells to ensure robust and consistent performance. These MEFs have been used successfully with ESC/iPSC lines from other species, but should be tested for compatibility if used for such an application.
Rely on a widely-used product
These CF1 MEFs are used in many labs worldwide and have been cited in various publications. They have been successfully used to support undifferentiated pluripotent stem cells at densities ranging from 2 x 104–5.3 x 104 cells/cm2.
For Research Use Only. Not for use in diagnostic procedures.
사양
공여자 소스Pooled
수량2 x 106 cells
배송 조건Shipped on dry ice
AgeEmbryonic
셀 유형Feeder Cells (Mouse Embryonic Fibroblast)
형태Cryopreserved
Mouse
Unit SizeEach
구성 및 보관
Cryopreserved mouse embryonic fibroblasts
Store in liquid nitrogen.
자주 묻는 질문(FAQ)
What is the difference between Mouse (ICR) Inactivated Embryonic Fibroblasts (Cat. No. A24903) and CF1 Mouse Embryonic Fibroblasts, irradiated (Cat. Nos. A34180, A34181)? Can they be used to reprogram fibroblasts?
Mouse (ICR) Inactivated Embryonic Fibroblasts and CF1 Mouse Embryonic Fibroblasts, irradiated are inactivated feeder layer fibroblast cells that are isolated from different mouse strains. Researchers often have a preference for one or the other. Neither of these can be used to reprogram fibroblasts.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Is it better to use irradiated mouse embryonic fibroblasts (MEFs) or Mitomycin C-treated MEFs to grow embryonic stem cells?
Either method will work in arresting cell division. However, the irradiation process will ensure that cell division will cease regardless of cell aggregation. Cell clumping can potentially not inactivate all cells when using mitomycin C, as cells within clumps may not be exposed to the mitomycin C. Irradiated cells are preferred by those who have concerns about chemical treatment. Mitomycin C-treated cells are preferred by those who have concerns about DNA damage from irradiation.
What are the differences between CF1, CF6, and DR4 mouse embryonic fibroblasts?
CF1 mouse embryonic fibroblasts do not have drug resistance. CF6 mouse embryonic fibroblasts are resistant to Neomycin/geneticin (G418). DR 4 mouse embryonic fibroblasts are resistant to geneticin (G418), puromycin, hygromycin, and 6-thioguanine.
What is the recommended seeding density for mouse embryonic fibroblasts? Why is the seeding density critical?
We recommend seeding these cells at densities ranging from 2 x 10E4 to 5.5 x 10E4 cells/cm2. A good starting point is 3 x 10E4 cells/cm2. If the feeder cells are too sparse, they may not maintain the pluripotent cells without differentiation, and the pluripotent cells may not attach well. If the feeder cells are too dense, the feeder layer may detach from the plate, and the culture will be lost.
When should mouse embryonic fibroblasts be plated and how long can these cells be used?
These cells should be plated 24 hours prior to plating the ESCs or iPSCs and should be used for only 7-10 days.