TrueGuide™ sgRNA Positive Control, HPRT1 (human) - FAQs
TrueGuide™ sgRNA Positive Control, HPRT1 (human) - FAQs
View additional product information for TrueGuide™ sgRNA Positive Control, HPRT1 (human) - FAQs (A35524)
1 product FAQs found
I would like to calculate the molecular weight of TrueGuide Synthetic gRNA? What is the length of TrueGuide Synthetic gRNA (TrueGuide sgRNA)?
The length of TrueGuide sgRNA is typically 100 nt. Also, the exact molecular weight can also be found on the COA.