Oncomine™ Focus Assay, AmpliSeq™ Library, 16 Samples - FAQs
Oncomine™ Focus Assay, AmpliSeq™ Library, 16 Samples - FAQs
View additional product information for Oncomine™ Focus Assay, AmpliSeq™ Library - FAQs (A29230, A35957)
1 product FAQs found
How many samples can I process with the Oncomine Focus Assay, Chef-Ready Library?
The Oncomine Focus Assay, Chef-Ready Library contains the following components needed to prepare libraries for 32 samples:
- Oncomine Focus Assay, DNA Chef-Ready Panel (2X) - contains the DNA primer pool
- Oncomine Focus Assay, RNA Chef-Ready Panel (2X) - contains the RNA primer pool
- Ion AmpliSeq Kit for Chef Reagents DL8 cartridge - allows automation of the library prep on the Ion Chef Instrument
Note: The SuperScript transcriptase for cDNA synthesis is not included and must be purchased separately.