Amplex™ UltraRed Reagent, 5 x 1 mg - FAQs
Amplex™ UltraRed Reagent, 5 x 1 mg - FAQs
View additional product information for Amplex™ Red and Amplex™ UltraRed Reagents - FAQs (A36006, A22177, A12222)
1 product FAQs found
I'm using an Amplex Red kit, the reagent changes color to pink almost immediately in my own Krebs-Ringer buffer but not in HBSS. Why is this?
The components of Krebs-Ringer buffer (salts) should not cause oxidation of the Amplex reagent (which, in the presence of peroxidase and H2O2 oxidizes to resorufin, which is pink in color and fluorescent). Try water alone (the water used to make the Krebs-Ringer buffer). Since Hank's Buffered Saline Solution is typically purchased rather than made in the lab, it likely would not have the same contaminant. Another option is to degas the buffer prior to use to removed dissolved oxygen radicals.