Veriti™ Thermal Cycler Extended Warranty + Plastics Package, REX, 384-well - FAQs
Veriti™ Thermal Cycler Extended Warranty + Plastics Package, REX, 384-well - FAQs
View additional product information for Veriti™ Thermal Cycler Extended Warranty + Plastics Package, REX, 384-well - FAQs (A36965)
1 product FAQs found
What is the optimal ramp rate for my thermal cycler?
There is no universal ramp rate for a thermal cycler because both the block and sample ramp rates are different for each thermal cycler. The sample ramp rate changes with the volume of the sample since more time and energy are required to heat higher sample volumes. The ramp rate may also need to be changed for a particular PCR assay or primer pair if you are trying to use the same PCR assay on two different thermal cyclers, for example.
As a reference, the block ramp rate commonly used with DreamTaq Polymerase on the MiniAmp thermal cycler is 3.0 C/sec. In a 50 µL PCR reaction, this translates to a sample ramp rate of ~1 C/sec.
Please refer to the following application note for further information about thermocycler ramp rates: https://assets.thermofisher.com/TFS-Assets/LSG/brochures/Thermal_Cycler_Ramp_RatesAppNote.pdf
Find additional tips, troubleshooting help, and resources within ourPCR Thermal Cyclers Support Center.