Oncomine™ Pan-Cancer Cell-Free Assay - FAQs
Oncomine™ Pan-Cancer Cell-Free Assay - FAQs
View additional product information for Oncomine™ Pan-Cancer Cell-Free Assay - FAQs (A37664)
5 product FAQs found
What software can I use to analyze the Oncomine Pan-Cancer Cell-Free Assay?
Analysis of Oncomine Pan-Cancer Cell-Free Assay data is performed using Ion Reporter Software.
How many genes are covered by the Oncomine Pan-Cancer Cell-Free Assay?
52 unique genes are covered by the assay.
What is the sensitivity for the Oncomine Pan-Cancer Cell-Free Assay?
The assay has a 0.1% limit of detection (LOD) with 20 ng of cell-free DNA input.
How many libraries can I multiplex with the Oncomine Pan-Cancer Cell-Free Assay?
1 library can be used on an Ion 530 chip, 4 libraries can be used on an Ion 540 chip, and 8 libraries can be used on an Ion 550 chip.
What DNA isolation methods are recommended for the Oncomine Pan-Cancer Cell-Free Assay?
We recommend using the MagMAX Cell-Free DNA Isolation Kit (Cat. No. A29319) or the MagMAX Cell-Free Total Nucleic Acid Isolation Kit (Cat. No. A36716).