A549 (+hIGF-1) Control Lysate (1 mg/mL) - FAQs
A549 (+hIGF-1) Control Lysate (1 mg/mL) - FAQs
View additional product information for A549 (+hIGF-1) Control Lysate (1 mg/mL) - FAQs (A37960)
1 product FAQs found
What are the characteristics of A549 (+hIGF-1) Control Lysate, and what is its main purpose?
A549 (+hIGF-1) Control Lysate is a stimulated A549 cell lysate that has been validated for the expression levels and phosphorylation of key targets (AKT, PTEN, IRS1, IGF1-R, GSK3a, GSK3b, PRAS40, mTOR, p70S6K, and TSC2) in the AKT signaling pathway. It serves as a positive control for use with SureQuant AKT panel kits.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.