What are the characteristics of HCT 116 (+hIGF-1) Control Lysate, and what is its main purpose?
HCT 116 (+hIGF-1) Control Lysate is a stimulated HCT 116 cell lysate that has been validated for the expression levels and phosphorylation of key targets (NRAS, KRAS, and HRAS) in the RAS signaling pathway. It serves as a positive control for use with SureQuant RAS panel kits.