Collibri™ PS DNA Library Prep Kit for Illumina Systems, with UD indexes (Set A, 1-24), 24 preps - FAQs
Collibri™ PS DNA Library Prep Kit for Illumina Systems, with UD indexes (Set A, 1-24), 24 preps - FAQs
View additional product information for Collibri™ PS DNA Library Prep Kits for Illumina Systems - FAQs (A43612024, A43613024, A38614196, A38614096, A38614096W, A43611024, A38612024W, A38612024, A38613024)
1 product FAQs found
Can indexed adaptors or other modules be purchased separately for the Collibri DNA Library Prep kits?
Indexed adapters and other modules in the kit can be purchased separately. Please refer to thermofisher.com/collibriDNA and contact us for more details.