TaqMan™ Universal DNA Spike-In Control (5X) - FAQs
View additional product information for TaqMan™ Universal DNA Spike-In Control (5X) - FAQs (A39176)
1 product FAQs found
How can I check for the presence of inhibitors in my samples when running microbiota profiling experiments?
We offer a TaqMan Universal DNA Spike-In Control (Cat. No. A39175 or A39176) that can be added to samples before extraction, to monitor recovery efficiency.