TaqMan™ Universal RNA Spike In/Reverse Transcription (Xeno) Control
Applied Biosystems™

TaqMan™ Universal RNA Spike In/Reverse Transcription (Xeno) Control

The TaqMan Universal RNA Spike In/Reverse Transcription (Xeno) Control is a synthetic RNA control that serves as an exogenous processRead more
Have Questions?
Catalog NumberQuantity
A391795 x 200 μL
Catalog number A39179
Price (USD)
864.86
Special offer
Online exclusive
Ends: 11-Dec-2026
1,042.00
Save 177.14
Each
Quantity:
5 x 200 μL
Price (USD)
864.86
Special offer
Online exclusive
Ends: 11-Dec-2026
1,042.00
Save 177.14
Each
The TaqMan Universal RNA Spike In/Reverse Transcription (Xeno) Control is a synthetic RNA control that serves as an exogenous process control for nucleic acid isolation and RNA recovery, reverse transcription, pre-amplification, and PCR. The control is used with the proprietary TaqMan assay for Xeno sequences (Assay IDs Ac00010014_a1 or Pa00010014_a1).

The TaqMan Universal RNA Spike In/Reverse Transcription (Xeno) Control is supplied at a concentration of 10,000 copies/μL. During nucleic acid isolation, 10 μL of the control is added to each test sample along with the nucleic acid binding reagents. The control is used to identify sample-specific amplification inhibition, reducing the likelihood of false negatives and providing confidence that results are accurate.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
FormatTube
Product LineTaqMan
Quantity5 x 200 μL
Shipping ConditionDry ice
Volume1000 μL
Concentration10,000 Copies/μL
Detection MethodPrimer-probe
For Use With (Application)Pathogen Detection
Unit SizeEach
Contents & Storage
5 tubes containing 200 μL

Store at -15° to -25°C.