TaqMan™ Universal Extraction Control Organism
Applied Biosystems™

TaqMan™ Universal Extraction Control Organism

The TaqMan Universal Extraction Control Organism contains three vials of lyophilized Bacillus atrophaeus for use as a process control forRead more
Have Questions?
Catalog NumberQuantity
A391803 pellets
Catalog number A39180
Price (USD)
473.00
Each
Quantity:
3 pellets
Price (USD)
473.00
Each
The TaqMan Universal Extraction Control Organism contains three vials of lyophilized Bacillus atrophaeus for use as a process control for cell lysis and nucleic acid recovery. This control is used with the proprietary TaqMan assay for Bacillus atrophaeus (Assay ID Ba06596576_s1). Like other gram-positive bacteria, Bacillus atrophaeus has thick cell walls than can be difficult to lyse. This characteristic makes gram-positive bacteria an ideal control to monitor the efficiency of cell lysis and subsequent nucleic acid recovery.

TaqMan Universal Extraction Control Organism is supplied lyophilized in a quantity of 1 × 109 copies/vial and is reconstituted in 200 μL of 1X PBS, pH 7.4 to a final concentration 5 × 16 copies/μL. During nucleic acid isolation, 10 μL of the control is processed as a stand-alone sample in a background of universal transport media. It can be added to the negative extraction control, and may also be added to one or more test samples at the start of the extraction process. The control is carried through the remainder of the workflow with the test samples. It is recommended that at least one stand-alone control sample is run per extraction plate.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
FormatVial
No. of Reactions60 Reactions
Product LineTaqMan
Quantity3 pellets
Shipping ConditionRoom temperature
Concentration1 x 109 Copies/Vial
Detection MethodPrimer-probe
For Use With (Application)Pathogen Detection
Unit SizeEach
Contents & Storage
3 vials containing lyophilized Bacillus atrophaeus.

Store at 4°C.

Frequently asked questions (FAQs)

Does the TaqMan Universal Extraction Control Organism (B. atrophaeus) (Cat No. A39180) contain inactivated or viable Bacillus atrophaeus?

TaqMan Universal Extraction Control Organism contain viable bacteria that can be cultured.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Applications Support Center.