TaqMan™ Universal Extraction Control Organism - FAQs
TaqMan™ Universal Extraction Control Organism - FAQs
View additional product information for TaqMan™ Universal Extraction Control Organism - FAQs (A39180)
1 product FAQs found
Does the TaqMan Universal Extraction Control Organism (B. atrophaeus) (Cat No. A39180) contain inactivated or viable Bacillus atrophaeus?
TaqMan Universal Extraction Control Organism contain viable bacteria that can be cultured.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Applications Support Center.