Pierce™ Chloroacetamide, No-Weigh™ Format, 10 x 2 mg - FAQs
Pierce™ Chloroacetamide, No-Weigh™ Format, 10 x 2 mg - FAQs
View additional product information for Pierce™ Alkylating Reagents - FAQs (A39270, A39271)
1 product FAQs found
How does a product with "No-Weigh" format reduce my concern over reagent stability?
The No-Weigh product format, where small amounts of reagent are pre-weighed and packaged separately, prevents the contamination and loss of reagent reactivity over time associated with repetitive opening and closing of a single vial. This format enables use of a fresh vial of reagent each time and eliminates the hassle of weighing out small amounts of reagent. Please view additional information and the video at: https://www.thermofisher.com/us/en/home/life-science/protein-biology/protein-labeling-crosslinking/no-weigh-packaging.html
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.