KingFisher™ Duo Prime Purification System with SmartStart
Thermo Scientific™
KingFisher™ Duo Prime Purification System with SmartStart
Reap high yields of quality nucleic acids from a variety of starting materials with the Thermo Scientific KingFisher Duo PrimeRead more
Have Questions?
Catalog NumberQuantity
A399561 package
Catalog number A39956
Price (USD)
39,275.81
Each
Quantity:
1 package
Price (USD)
39,275.81
Each
Reap high yields of quality nucleic acids from a variety of starting materials with the Thermo Scientific KingFisher Duo Prime Purification System. This compact instrument is ideal for laboratories requiring nucleic acid purification and is easily modified for use in proteomics and cell isolation. It provides excellent reproducibility with minimal hands-on time and has a small footprint in the lab. A UV lamp for easy decontamination and optional bar-code reader enhance its functionality.
This package includes the KingFisher Duo Prime Purification System and SmartSmart Orientation.
Features of this KingFisher Duo Prime system package include:
• DNA, RNA, and protein isolation from a variety of starting materials, including cell-free body fluids, blood, bacteria, cell culture, tissue, and plant samples, with volumes ranging up to 5 mL
• Automated magnetic bead technology allows excellent recovery of samples with great reproducibility and efficiency
• Optional bar-code reader allows tracking of samples in the internal software
• Optimized for use with KingFisher Pure DNA and RNA isolation kits and protocols for a complete, easy-to-use solution, or customize with BindIt Software to create your own protocols
• 1-day SmartStart Orientation to get you up and running quickly
More about the KingFisher Duo Prime Purification System ›
Get up and running fast with SmartStart Orientation
The SmartStart Orientation included in this package is performed by a Field Application Scientist (FAS) in your lab and helps you:
• Get up and running quickly
• Get connected to your local network and Cloud
• Choose appropriate reagents and consumables to meet your needs
Content will vary depending on your level of knowledge and experience.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Customer Education1 day at customer site
DescriptionKingFisher™ Duo Prime Purification System
For Use With (Application)DNA and RNA Isolation From Various Starting Materials, Proteomic Applications, Cell Isolation
For Use With (Equipment)Nucleic Acid Purification Systems
IncludesKingFisher™ Duo Prime Purification System and SmartStart 1 Day Training
Particle Size (Metric)ca. >1 μm
Quantity1 package
Total Service Coverage1 year manufacturer warranty only
Volume (Metric) Processing96 DW plate 50 to 1000 μL, 24 DW plate 200 to 5000 μL, Elution strip 30 to 130 μL
Unit SizeEach
Package Components
KingFisher™ Duo Prime Purification System
Catalog Number
5400110
Unit Size
Each
Qty
1
TRN00340
Catalog Number
TRN00340
Unit Size
Qty
1
Frequently asked questions (FAQs)
Where can I find the manual for the BindIt Software?
The User Manual for the BindIt Software can be downloaded from the BindIt Software product page online, under the Documents & Downloads section. It is also included within the BindIt Software installation file under "Documentation".
Find additional tips, troubleshooting help, and resources within our Automated Nucleic Acid Purification Support Center.
Do you have data that shows how the UV lamp of the KingFisher Duo Prime instrument gets rid of bacterial contamination?
Yes we have an application note that demonstrates the lethal effect of UVC irradiation on bacterial growth and deactivation of human gDNA, "Decontamination of the KingFisher Duo Prime processing chamber using UV light".
Find additional tips, troubleshooting help, and resources within our Automated Nucleic Acid Purification Support Center.
What type of tip heads do I receive when I purchase the KingFisher Duo Prime instrument?
You will receive the following tip heads when you purchase a KingFisher Duo Prime instrument:
• 6-tip head (for 24 deep-well plate)
• 12-tip head (for 96 deep-well plate)
Find additional tips, troubleshooting help, and resources within our Nucleic Acid Purification and Analysis Support Center.
Do you sell elution strips for the KingFisher Duo Prime 24 Deep-Well (6-tip) format?
We do not sell elution strips for the KingFisher Duo Prime 24 Deep-Well format. A 24 Deep-Well plate must be used for elution with the 24 Deep-Well protocol.
Find additional tips, troubleshooting help, and resources within our Nucleic Acid Purification and Analysis Support Center.
Are the Matrix 500 microliter ScrewTop Tubes (Cat. No. 3743AMB) compatible with the Kingfisher Duo Prime barcode reader?
Yes, the KingFisher Duo Prime barcode reader is able to read the 2D barcode on the Matrix 500 µL ScrewTop Tubes. The barcode reader can generate an error if the tubes are in the tube rack and the barcode position is not able to be scanned. If this occurs, we recommend scanning each tube individually.
Find additional tips, troubleshooting help, and resources within our Automated Nucleic Acid Purification Support Center.