TMTpro™ 10plex Isobaric Label Reagent Set (Unit Mass Reporter), 1 x 5 mg
Thermo Scientific™
TMTpro™ 10plex Isobaric Label Reagent Set (Unit Mass Reporter), 1 x 5 mg
The Thermo Scientific™ TMTpro 10plex Label Reagent Set (Unit Mass Reporter), 1 x 5 mg enables multiplexing of up to 10 samples for simultaneous analysis on lower resolution MS instruments.
Have Questions?
Catalog NumberQuantity
A400009281 x 5 mg
Catalog number A40000928
Price (USD)
6,430.00
Each
Quantity:
1 x 5 mg
Price (USD)
6,430.00
Each
The Thermo Scientific™ TMTpro 10plex Label Reagent Set (Unit Mass Reporter), 1 x 5 mg enables multiplexing of up to 10 samples for simultaneous analysis on lower resolution MS instruments.
For Research Use Only.
Specifications
Final Product Typepeptide
For Use With (Equipment)Mass Spectrometer
Quantity1 x 5 mg
Workflow StepPeptide labeling
Detection MethodMass Spectrometry, Mass Spectrometry
FormPowder
Product LineTMTpro
Product Type32plex Label Reagent Matched Set
Starting MaterialPeptides
Unit SizeEach
Frequently asked questions (FAQs)
What are the primary applications of the TMTpro 10plex Isobaric Label Reagent Set (Unit Mass Reporter), 1 x 5 mg (Cat. No. A40000928)?
The TMTpro 10plex Isobaric Label Reagent Set (Unit Mass Reporter), 1 x 5 mg (Cat. No. A40000928) enables high-precision quantitative proteomics across a broad range of MS platforms. It features reagents with a 1 Da mass difference between reporter ions, making it ideal for use on non-Orbitrap instruments with low MS/MS resolving power, such as the Thermo Scientific Stellar series MS instruments and various MS instruments from other manufacturers.
This reagent set also supports 10plex quantification using the complementary ion cluster in MS2 spectra, which can enhance quantification accuracy and precision on Orbitrap and non-Orbitrap platforms. For more detailed information, please refer to the following publications:
https://pubs.acs.org/doi/10.1021/acs.jproteome.0c00813
https://pubmed.ncbi.nlm.nih.gov/39725028/
Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.
What is the shelf life for the TMTpro 32plex and TMTpro Deuterated Label Reagents?
The TMTpro 32plex and TMTpro Deuterated Label Reagents are guaranteed for one year from the date of receipt when stored at the recommended temperature in original, unopened packaging.
Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.
If I already have TMTpro 16plex Label Reagent Set (Cat. No. A44520), what do I need to purchase to perform 32plex experiments?
You would need to use TMTpro 16plex Label Reagent Set (Cat. No. A44520) in combination with TMTpro 16plex Deuterated Label Reagent Set (Cat. No. A40000817).
Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.
Can I purchase the TMTpro-134C and TMTpro-135CD Label Reagents separately from the TMTpro 32plex and TMTpro Deuterated Label Reagents?
Yes, 134C and 135CD Label Reagents (Cat. No. A40000853) provides TMTpro-134C and TMTpro-135CD tags in two separate vials with each vial containing 5 mg of the tag in lyophilized format. 135CD Label Reagent (Cat. No. A40000818) provides one vial containing 5 mg of TMTpro-135 CD tag in lyophilized format.
Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.
What is provided with the 134C and 135CD Label Reagents (Cat. No. A40000853)?
134C and 135CD Label Reagents (Cat. No. A40000853) provides two TMTpro tags, TMTpro-134C and TMTpro-135CD. The tags are supplied in two separate vials, with each vial containing 5 mg of the tag in lyophilized format.
Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.