Absolute Q™ MAP16 Plate Kit and Absolute Q™ Universal DNA dPCR Master Mix
Applied Biosystems™

Absolute Q™ MAP16 Plate Kit and Absolute Q™ Universal DNA dPCR Master Mix

Eliminate the need to design custom assays and choose from over 20 million predesigned assays. Includes the Absolute Q MAP16 plate kit plus the Absolute Q Universal DNA dPCR master mix which is compatible with a wide range of applications and difficult targets.
Have Questions?
Catalog NumberQuantity
A400016971 Ea.
Catalog number A40001697
Price (MXN)
-
Quantity:
1 Ea.
The Absolute Q MAP16 Plate Kit and Absolute Q Universal DNA Digital PCR Master Mix are key components of the QuantStudio Absolute Q Digital PCR System. The system is powered by microfluidic array plate (MAP) technology that enables consistent and automated separation of the sample into 20,000 microchambers with industry leading <1% coefficient of variation (CV).

Absolute Q MAP16 Plate Kit

The Absolute Q MAP16 microinjection-molded plates overcome common challenges of inconsistency and high dead volume. This novel approach provides many benefits, including precise microchamber volume, consistent sample distribution, and greater than 98% of loaded volume analyzed.

Absolute Q Universal DNA Digital PCR Master Mix

This package product also includes Absolute Q Universal DNA Digital PCR Master Mix, which is compatible with a broad range of applications and over 20 million predesigned TaqMan Assays (performance-guaranteed*), reducing the need to design custom assays. Simplify your workflow by avoiding assay design and minimizing assay optimization; use the same assay for both qPCR and digital PCR. In addition, with 72 hours of assembled-reaction benchtop stability, this master mix enables increased productivity through automation.

The impressive precision and accuracy of the Absolute Q system helps provide confidence in measurements across replicates and experiments.

* We guarantee that our predesigned TaqMan Assays will enable you to obtain data you can trust, or we'll replace them at no cost or give you a credit for the assay purchase price. Terms and conditions apply.

For Research Use Only.
Specifications
No. of Reactions192 Reactions
Quantity1 Ea.
Detection MethodPrimer-probe
For Use With (Application)Digital PCR (dPCR), DNA Quantitation, Gene Expression, Genotyping, Microbial Detection
FormLiquid
GC-Rich PCR PerformanceHigh
PCR MethodDigital PCR
Reaction SpeedFast or Standard
Unit SizeEach
Contents & Storage
12 MAP16 Plates, Gaskets, Isolation Buffer, and 1 tube Absolute Q™ Universal DNA Digital PCR Master Mix (192 reactions)

Frequently asked questions (FAQs)

How many MAP16 plates are included in the Absolute Q MAP16 Plate Kit and Absolute Q Universal DNA dPCR Master Mix (Cat. No. A40001697)

12 packs of MAP16 plates.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Applications Support Center.