Dry-Ready™ RT-LAMP Kit
Invitrogen™

Dry-Ready™ RT-LAMP Kit

Experience the simplicity and efficiency of the air drying process by gently heating materials at moderate temperatures in a simple ventilated oven.
Have Questions?
Catalog NumberQuantity
A40006907200 Reactions
Catalog number A40006907
Price (USD)
294.00
Each
Quantity:
200 Reactions
Request bulk or custom format
Price (USD)
294.00
Each

The Dry-Ready RT-LAMP Kit is a complete solution for reverse transcription loop-mediated isothermal amplification (RT-LAMP)-based assay air drying. The kit includes all required components: Dry-Ready SuperScript IV Reverse Transcriptase, Dry-Ready Bst DNA Polymerase, Dry-Ready dNTP Mix, and 10X Dry-Ready RT-LAMP Reaction Buffer to facilitate pre- and post-drying procedures.

Features of the Dry-Ready RT-LAMP Kit include:

  • All required components for RT-LAMP assay drying and downstream applications
  • Rapid RNA conversion in RT-LAMP reaction using SuperScript IV Reverse Transcriptase
  • Robust Bst DNA Polymerase with enhanced speed and sensitivity, engineered for fast and efficient amplification in RT-LAMP

The Dry-Ready RT-LAMP Kit includes an evolved Bst DNA polymerase mutant with a fast reaction speed and increased sensitivity and tolerance to inhibitors, as well as SuperScript IV Reverse Transcriptase with increased processivity, thermostability, and inhibitor resistance.

The Dry-Ready kit is optimized for the air drying process. Notably, air drying is a simple process that does not impair functionality of the assay. On the contrary, it help provides flexibility on the initial volume of sample and increases sensitivity, resulting in a exceptional stabilization process for point-of-care applications.

For Research Use Only.
Specifications
No. of Reactions200 Reactions
Optimal Reaction Temperature65°C
Reaction Time≤10 Min.
Reverse TranscriptaseSuperScript™ IV Reverse Transcriptase
For Use With (Application)RT-LAMP, LAMP
FormLiquid
PolymeraseBst DNA Polymerase
Product TypeDNA Amplification Kit
Quantity200 Reactions
Research CategoryNucleic Acid Amplification
Unit SizeEach
Contents & Storage

•Dry-Ready™ Bst DNA Polymerase
•Dry-Ready™ SuperScript™ IV Reverse Transcriptase
•Dry-Ready™ dNTP Mix
•5X Dry-Ready™ Reaction Buffer
•MgCl2 Solution
•Store at -15 to -25°C