Recombinant Bovine Serum Albumin (rBSA), AOF
Invitrogen™

Recombinant Bovine Serum Albumin (rBSA), AOF

For consistent performance in a variety of molecular biology applications that require animal-free formulations, high quality recombinant BSA can be used to stabilize enzymes, enhance enzymatic activity, or to block non-specific binding.
Have Questions?
Catalog NumberQuantity
A400074905 mg
Catalog number A40007490
Price (USD)
24.50
Each
Quantity:
5 mg
Price (USD)
24.50
Each

Invitrogen Recombinant Bovine Serum Albumin (rBSA), AOF, is designed to deliver exceptional quality and consistent performance for a variety of molecular biology applications that require animal origin-free formulations. Recombinant BSA manufacturing technology helps ensure simplified logistics, regulatory compliance, and protection from animal related illnesses.

Recombinant BSA is a protein used to stabilize enzymes during long-term storage and prolonged enzymatic reactions. The presence of rBSA can enhance enzymatic efficiency in the presence of inhibitors (such as during PCR) and prevent the adhesion of enzymes to reaction tubes and tip surfaces.

Features of Invitrogen rBSA, AOF, include:

  • Expressed in yeast—helps ensure correct protein folding and stability
  • Animal origin-free (AOF)—suitable for applications requiring non-animal derived components
  • RNase, DNase, and protease-free—helps prevent degradation of nucleic acids and proteins

Applications of Invitrogen rBSA, AOF, include:

  • Enzyme stabilization—helps maintain enzyme activity during storage and prolonged enzymatic reactions
  • PCR and RT-qPCR—helps protects DNA polymerases and reverse transcriptases from sample-based inhibition, resulting in enhanced performance
  • DNA digestion with restriction endonucleases—helps ensure enzyme stability during restriction digestion reaction
  • ELISA—helps prevent non-specific binding and improve assay specificity
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product TypeRecombinant BSA
ProteinBovine Serum Albumin
Purity≥95%
Quantity5 mg
SourcePichia pastoris
Concentration20 mg/mL
FormSolution
Research CategoryTraditional Cloning, Scarless Cloning, PCR
Sensitive to Heat InactivationYes
Unit SizeEach