Genexus™ Controls
Ion Torrent™

Genexus™ Controls

The Ion Torrent Genexus Controls kit provides library and template preparation controls for use with the Ion Torrent Genexus Integrated Sequencer.
Have Questions?
Catalog NumberQuantity
A402671 kit
Catalog number A40267
Price (USD)
1,764.00
Each
Quantity:
1 kit
Price (USD)
1,764.00
Each

The Ion Torrent Genexus Controls kit provides library and template preparation controls for use with the Ion Torrent Genexus Integrated Sequencer. The kit offers greater experimental control and empowers you to assess sample quality, help save time and reagents, and maximize productivity. Each kit contains an Ion AmpliSeq-based Genexus Control Panel and a Genexus Control DNA, which can prepare up to 32 automated control libraries, as well as a Genexus Control Library, sufficient for four template preparations.

The Ion Torrent Genexus Integrated Sequencer automates library preparation, sequencing, analysis, and reporting in a hands-off, walkaway workflow to deliver results in a single day. The innovative, multilane sequencing chip is designed to enable cost-effective processing of 1 to 32 single pool libraries.

Related Links:

For Research Use Only. Not for use in diagnostic procedures.

Specifications
For Use With (Application)Sequencing
For Use With (Equipment)Genexus™ Integrated Sequencer
LibrariesTargeted Sequencing Library
Product LineGenexus
Product TypeControl
Quantity1 kit
Sample TypeDNA, RNA
Shipping ConditionDry Ice
Workflow StepLibrary Generation
Sequencing TypeGenome, DNA, and RNA Sequencing
Unit SizeEach
Contents & Storage
Sufficient for 32 DNA to sequencing reactions OR 4 library to sequencing reactions

Store at -10 to -30°C.

Frequently asked questions (FAQs)

What is the level of detection for somatic variants with the Oncomine Myeloid Research Assay GX?

Detection of somatic variants with the Oncomine Myeloid Research Assay GX has been verified down to 5% allele frequency.

Which extraction kits are compatible with Oncomine Myeloid Research Assay GX?

Here is a list of kits that are compatible with Oncomine Myeloid Research Assay GX:

Nucleic acid isolation - RNA samples
- MagMAX mirVana Total RNA Isolation Kit, Cat. No. A27828
- PureLink Total RNA Blood Kit, Cat. No. K156001
- RNaseZap RNase Decontamination Solution, Cat. No. AM9780, AM9782, AM9784

Nucleic acid isolation - DNA samples
- MagMAX DNA Multi-Sample Ultra 2.0 Kit, Cat. No. A36570
- PureLink Genomic DNA Mini Kit, Cat. No. K1820-01

What are the sample input requirements for Oncomine Myeloid Research Assay GX?

The recommended minimum sample input is 27.75 ng of DNA (1.11 ng/µL minimum concentration) and 14.25 ng of RNA (0.95 ng/µL minimum concentration).

What sample controls should I use with the Oncomine Myeloid Research Assay GX?

The recommended controls are:

DNA Controls:
- Seraseq® Myeloid Mutation DNA Mix, Cat. No. 0710-0408 (SeraCare®)
- Myeloid DNA Reference Standard, Cat. No. HD829 (Horizon™)
- AcroMetrix™ Oncology Hotspot Control, Cat. No. 969056

RNA Controls:
- Universal Human Reference RNA, Cat. No. 740000 (Agilent™)
- Seraseq® Myeloid Fusion RNA Mix, Cat. No. 0710-0407 (SeraCare®)

How many Oncomine Myeloid Research Assay GX samples can be multiplexed on a GX5 Chip?

Multiplex sequencing of up to 8 Oncomine Myeloid Research Assay GX samples (8 DNA+ 8 RNA) per lane on a GX5 Chip can be performed in a single run. There are 4 lanes on the GX5 Chip, so 32 total samples (32 DNA+ 32 RNA) can be run on one GX5 Chip (8 samples per run).