Genexus™ GX5™ Starter Pack-HD - FAQs
Genexus™ GX5™ Starter Pack-HD - FAQs
View additional product information for Genexus™ GX5™ Starter Pack-HD - FAQs (A40280)
2 product FAQs found
What are the correct shipping conditions for the Genexus Cartridges that are a part of the Genexus Sequencing Kit (Cat. No. A40271) and Genexus GX5 Starter Packs (Cat. Nos. A40279, A40280)?
The Genexus Cartridge should be shipped on supergels at -15 to -50 degrees C, in a polystyrene box. If the cartridges are shipped with dry ice or arrive thawed, they should not be used, and replacement would need to be issued.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
What are the components of the Genexus GX5 Starter Pack-HD?
Genexus GX5 Starter Pack-HD (Cat. No. A40280) supplies the following components for Ion AmpliSeq HD library preparation and sequencing using a Genexus-ready assay:
- Genexus Strip 1, 8 strips
- Genexus Strip 2‑HD, 8 strips
- Genexus Strip 3‑GX5, 8 strips
- Genexus Strip 4, 8 strips
- Genexus Barcodes 1-32 HD, 1 plate
- Genexus Pipette Tips, 12 racks
- Genexus Cartridge, 2 cartridges
- Genexus Bottle 2, 4 bottles
- Genexus Bottles 1 and 3, 2 bottles each
Note:
- Genexus Barcodes 1-32 HD (Cat. No. A40261) and Genexus Pipette Tips (Cat. No. A40266) are also orderable individually.
- Library and template strips are orderable separately in paired sets as Genexus Library Strips 1 and 2-HD (Cat. No. A40255) and Genexus Templating Strips 3 and 4-GX5 (Cat. No. A40263).
- The Genexus Cartridge, Genexus Bottle 2, and Genexus Bottles 1 and 3 are orderable as a set in the Genexus Sequencing Kit (Cat. No. A40271).
- For custom assays, Genexus Primer Pool Tubes (Cat. No. A40262) must be ordered separately.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.