TaqMan™ Arbovirus Triplex Control Kit (ZIKV/DENV/CHIKV)
TaqMan™ Arbovirus Triplex Control Kit (ZIKV/DENV/CHIKV)
TaqMan™ Arbovirus Triplex Control Kit (ZIKV/DENV/CHIKV)
TaqMan™ Arbovirus Triplex Control Kit (ZIKV/DENV/CHIKV)
Applied Biosystems™

TaqMan™ Arbovirus Triplex Control Kit (ZIKV/DENV/CHIKV)

The Applied Biosystems TaqMan Arbovirus Triplex Control Kit (ZIKV/DENV/CHIKV) contains human DNA (1X) and pooled complete genome RNA template ofRead more
Have Questions?
Catalog NumberNo. of Reactions
A4049025 Reactions
Catalog number A40490
Price (USD)
784.00
Each
Add to cart
No. of Reactions:
25 Reactions
Price (USD)
784.00
Each
Add to cart
The Applied Biosystems TaqMan Arbovirus Triplex Control Kit (ZIKV/DENV/CHIKV) contains human DNA (1X) and pooled complete genome RNA template of the following viruses:
• Zika virus with Asian lineage
• Dengue virus from four serotypes (DENV1, DENV2, DENV3, and DENV4)
• Chikungunya virus
 
This control kit is used with these detection kits:
• TaqMan Arbovirus Triplex Kit (ZIKV/DENV/CHIKV), 0.2 mL (Cat. No. A31746)
• TaqMan Arbovirus Triplex Kit (ZIKV/DENV/CHIKV), 0.1 mL (Cat. No. A31747)
 
Specifications
For Use WithTaqMan™ Zika Virus Triplex Kit
No. of Reactions25 Reactions
Product LineTaqMan
Shelf Life2 years from the date of manufacture
Shipping ConditionRoom Temperature
Unit SizeEach