TrueMark™ OpenArray™ Respiratory Tract Microbiota Plate
Applied Biosystems™
TrueMark™ OpenArray™ Respiratory Tract Microbiota Plate
TrueMark OpenArray Respiratory Tract Microbiota Plate is an efficient, easy-to-use OpenArray plate (112-assay format) for the characterization of key respiratoryRead more
Have Questions?
Catalog NumberQuantity
A412371 plate
Catalog number A41237
Price (USD)
1,628.00
Each
Quantity:
1 plate
Price (USD)
1,628.00
Each
TrueMark OpenArray Respiratory Tract Microbiota Plate is an efficient, easy-to-use OpenArray plate (112-assay format) for the characterization of key respiratory tract microbial targets through real-time PCR. This plate contains TaqMan Microbial Assays for 31 microbial targets, plus Universal Extraction Organism Control (B. atropheus), Universal DNA Spike-In Positive Control (Xeno) and human RNase P RPPH1 control. The format of this OpenArray plate allows for three replicates to be run in parallel per plate for all targets and two replicates for B. atropheus and RNase P. Each OpenArray can be used for running 23 samples and 1 control sample.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product TypeRespiratory Tract Microbiota Plate
Quantity1 plate
Shipping ConditionDry Ice
Detection MethodPrimer-probe
GC-Rich PCR PerformanceLow
Unit SizeEach
Contents & Storage
Store in freezer (-5 to -30°C).