Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Proteins & Peptides ›
          • CD64 Proteins

          Invitrogen

          Mouse Fc gamma RI (CD64) His-tag Recombinant Protein

          View all (4) CD64 proteins

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Tech Support
          Datasheet
          Certificates
          Tech Support
          Datasheet
          Certificates
          Tech Support

          Cite Mouse Fc gamma RI (CD64) His-tag Recombinant Protein

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation'] || 'Additional Information'}}: {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Testing Data (1)
          Mouse Fc gamma RI (CD64) His-tag Protein
          Group 53 Created with Sketch.
          Mouse Fc gamma RI (CD64) His-tag Protein
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Mouse Fc gamma RI (CD64) His-tag Protein (A42617)

          SDS-PAGE analysis of Mouse Fc gamma RI (CD64) His-tag (Product # A42617). The specification for purity shows: ≥ 95 % (SDS-PAGE), ≥ 90 % (SEC-HPLC). ≥ 90 % (SEC-MALS, routine testing). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Mouse Fc gamma RI (CD64) His-tag Protein

          Product Details

          A42617

          Applications
          Tested Dilution
          Publications

          Control (Ctrl)

          Assay-Dependent
          -

          Binding assay (Binding)

          Assay-dependent
          -
          Product Specifications

          Species

          Mouse

          Expression System

          HEK293 cells

          Amino acid sequence

          Mouse FCGR1, amino acids Met1-Pro297 (Accession # NP_034316.1) with a C-terminal His-tag

          Tag

          His-tag

          Molecular weight

          32 kDa

          Class

          Recombinant

          Type

          Protein

          Purity

          >95 % as determined by SDS-PAGE. >90 % as determined by SEC-HPLC

          Endotoxin concentration

          <1.0 EU/µg

          Activity

          1.Immobilized Rituximab (IgG1) at 1 µg/mL can bind Mouse Fc gamma RI (CD64) His-tag Recombinant Protein, the EC50 is 16-35 ng/mL.
          2.Immobilized Anti-CD3 antibody, mouse IgG2a at 2 µg/mL (100 µL/well) can bind Mouse Fc gamma RI (CD64) His-tag Recombinant Protein, the EC50 is 0.8-2.4 ng/mL(QC tested).
          3.Loaded Mouse Fc gamma RI (CD64) His-tag Recombinant Protein on His1K Biosensor, can bind Bevacizumab (IgG1) with an affinity constant of 0.12 µM as determined in a BLI assay (Sartorius Octet Red384) (Routinely tested).
          4.Loaded Mouse Fc gamma RI (CD64) His-tag Recombinant Protein on His1K Biosensor, can bind Nivolumab (IgG4) with an affinity constant of 0.28 µM as determined in a BLI assay (Sartorius Octet Red384) (Routinely tested).
          5.Captured Mouse Fc gamma RI (CD64) His-tag Recombinant Protein on Anti-His Chip can bind Bevacizumab (IgG1) with an affinity constant of 0.12 µM as determined in an SPR assay (Biacore T200) (Routinely tested).
          6.Captured Mouse Fc gamma RI (CD64) His-tag Recombinant Protein on Anti-His Chip can bind Nivolumab (IgG4) with an affinity constant of 0.69 µM as determined in an SPR assay (Biacore T200) (Routinely tested).
          7.The purity of Mouse Fc gamma RI (CD64) His-tag Recombinant Protein is more than 95% and the molecular weight of this protein is around 40-50 kDa verified by SEC-MALS.
          8.Loaded Monoclonal Anti-Human CD3 Antibody, Mouse IgG2a (OKT3) on ProA Biosensor, can bind Mouse Fc gamma RI (CD64) His-tag Recombinant Protein with an affinity constant of 3.46 nM as determined in BLI assay (Sartorius Octet RED384)(QC tested).

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Lyophilized

          Amount

          50 µg

          Purification

          purified

          Storage buffer

          PBS, pH 7.4, with 5% mannitol, 5% trehalose, 0.01% Tween 80

          Contains

          no preservative

          Storage conditions

          -20°C or -80°C if preferred

          Shipping conditions

          Ambient

          Product Specific Information

          Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2 mg/mL or lower. It is recommended that the protein be aliquoted and be used as soon as possible. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.

          Target Information

          CD64 (FCGR1A) encodes a protein that plays an important role in the immune response. CD64 is a high-affinity Fc-gamma receptor and is one of three related gene family members located on chromosome 1. CD64 plays an important role in the immune response and its dysfunction has been studied in cervical adenitis and tetrasomy 21.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: CD64 antigen; Fc-gamma receptor 1; fc-gamma RI; fc?r; FcgammaRI; FcgRI; fcRI; FLJ18345; high affinity IgG receptor; igG Fc receptor I; IgG high affinity Fc receptor; RP11-196G18.2; unnamed protein product

          View more View less

          Gene Aliases: CD64; FcgammaRI; IGGHAFC

          View more View less

          UniProt ID: (Mouse) P26151

          View more View less

          Entrez Gene ID: (Mouse) 14129

          View more View less

          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5d6d8bc979-86tfw:80/100.66.78.224:80.
          git-commit: e71199a02a6c1449b2ac9ded8316d5734e8aa2de
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.62.0-2026.11.45-1.0